missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA6 / Annexin A6 / Annexin6 Protein
A DNA sequence encoding the Homo sapiens (Human) ANXA6 / Annexin A6 / Annexin6, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_001146.2 |
---|---|
Gene ID (Entrez) | 309 |
Name | ANXA6 / Annexin A6 / Annexin6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044071
|
enQuireBio™
QP8334-ec-10ug |
10 μg | N/A | ||||||
16084071
|
enQuireBio™
QP8334-ec-50ug |
50 μg | N/A | ||||||
16034071
|
enQuireBio™
QP8334-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16064071
|
enQuireBio™
QP8334-ec-200ug |
200 μg | N/A | ||||||
16074071
|
enQuireBio™
QP8334-ec-500ug |
500 μg | N/A | ||||||
16054071
|
enQuireBio™
QP8334-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001146.2 | |
ANXA6 / Annexin A6 / Annexin6 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
309 | |
Research Use Only | |
ANXA6 | |
Recombinant Protein | |
E. coli | |
AKPAQGAKYR GSIHDFPGFD PNQDAEALYT AMKGFGSDKE AILDIITSRS NRQRQEVCQS YKSLYGKDLI ADLKYELTGK FERLIVGLMR PPAYCDAKEI KDAISGIGTD EKCLIEILAS RTNEQMHQLV AAYKDAYERD LEADIIGDTS GHFQKMLVVL LQGTREEDDV VSEDLVQQDV QDLYEAGELK WGTDEAQFIY ILGNRSKQHL RLVFDEYLKT TGKPIEASIR GELSGDFEKL MLAV | |
Greater than 90% as determined by SDS-PAGE. |