missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human Cadherin-17 His (C-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ NBP3-18199
This item is not returnable.
View return policy
Specifications
LYOPH | |
Lyophilized from a 0.22 um filtered solution of PBS, pH 7.4 | |
100 μg | |
<0.1 EU/μg of the protein by LAL method | |
CDH17 | |
Cancer | |
Cadherin-17 |
SDS-PAGE | |
1015 | |
Store at -20°C. | |
cadherin, cadherin 17, LI cadherin (liver-intestine), cadherin-16, cadherin-17, CDH16, FLJ26931, HPT1, HPT-1, HPT-1 cadherin, human intestinal peptide-associated transporter HPT-1, human peptide transporter 1, Intestinal peptide-associated transporter HPT-1, LI cadherin, LI-cadherin, Liver-intestine cadherin, MGC138218, MGC142024 | |
Reconstitute to a concentration of 0.1-0.5 mg/mL in sterile distilled water | |
>90%, by SDS-PAGE |