missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Carbonic Anhydrase XIV Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 2195-CA-010

Additional Details : Weight : 0.00970kg

Product Code. 18107820

  • 3146.79 DKK / 10µg
Estimated Shipment: 20-08-2024
to see stock.

Description

Description

The Recombinant Human Carbonic Anhydrase XIV Protein is derived from NS0. The Recombinant Human Carbonic Anhydrase XIV Protein has been validated for the following applications: Enzyme Activity.
Specifications

Specifications

Supplied as a 0.2μm filtered solution in Tris and NaCl.
32 kDa
10μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
23632
Carbonic Anhydrase XIV/CA14
Mouse myeloma cell line,NS0-derived human Carbonic Anhydrase XIV/CA14 protein Gly19-Met290,with a C-terminal 10-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers