missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human CD11c GST (N-Term) Protein

Recombinant Protein

4551.72 DKK - 5966.37 DKK

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 3687
Molecular Weight (g/mol) 37.84 kDa
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18310639
View Documents Promotion Details
Novus Biologicals™
H00003687-Q01-25ug
25 μg
5966.37 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
18325988
View Documents Promotion Details
Novus Biologicals™
H00003687-Q01-10ug
10 μg
4551.72 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, Protein Array, Immunoaffinity Purification
3687
Store at -80°C. Avoid freeze-thaw cycles.
ITGAX
>80% by SDS-PAGE and Coomassie blue staining
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
37.84 kDa
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
Dendritic Cell Markers, Signal Transduction
CD11c
Videos
SDS
Documents

Documents

Product Certifications