Learn More
R&D Systems™ Recombinant Human CD39L1/ENTPD2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 6087-EN-010
This item is not returnable.
View return policy
Description
The Recombinant Human CD39L1/ENTPD2 Protein is derived from CHO. The Recombinant Human CD39L1/ENTPD2 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2 μm filtered solution in Tris, NaCl, CaCl 2 and Glycerol. | |
48 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
954 | |
CD39L1/ENTPD2 | |
Chinese Hamster Ovary cell line,CHO-derived human CD39L1/ENTPD2 protein Thr29-Asp460,with a C-terminal 6-His tag | |
Monomer | |
Recombinant |
Product Content Correction