missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CXCL7/NAP-2 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
1825.00 DKK - 3510.00 DKK
Specifications
Gene ID (Entrez) | 5473 |
---|---|
Molecular Weight (g/mol) | 7.6 kDa |
Name | CXCL7/NAP-2 |
Source | E. coli-derived human CXCL7/NAP-2 protein Ala59-Asp128 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Formulation | Price | Quantity & Availability | ||||
18327237
|
R&D Systems™
393-NP-010 |
10 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
1825.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18330538
|
R&D Systems™
393-NP-010/CF |
10 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
1825.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18198590
|
R&D Systems™
393-NP-050 |
50 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. |
3510.00 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18147160
|
R&D Systems™
393-NP-050/CF |
50 μg | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. |
3510.00 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Human CXCL7/NAP-2 Protein is derived from E. coli. The Recombinant Human CXCL7/NAP-2 Protein has been validated for the following applications: Bioactivity.Specifications
5473 | |
CXCL7/NAP-2 | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
7.6 kDa | |
E. coli-derived human CXCL7/NAP-2 protein Ala59-Asp128 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |