missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA polymerase processivity factor Protein
A DNA sequence encoding the Homo sapiens (Human) DNA polymerase processivity factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9615-ye-50ug
This item is not returnable.
View return policy
Specifications
DNA polymerase processivity factor Protein | |
Research Use Only | |
U27 | |
Recombinant Protein | |
Yeast | |
FFFFKAHKAR VGARTSFLTE MERGSRDHHR DHRDHREHRE TREPPTLAFH MKSWKTINKS LKAFAKLLKE NATVTFTPQP SIIIQSAKNH LVQKLTIQAE CLFLSDTDRF LTKTINNHIP LFESFMNIIS NPEVTKMYIQ HDSDLYTRVL VTASDTCTQA SVPCVHGQEV VRDTGRSPLR IDLDHSTVSD VLKWLSPVTK TKRSGKSDAL MAHIIVQVNP PTIKFVTEMN ELEFSNSNKV IFYDVKNMRF NLSAKNLQQA LSMCAVIKTS CSLRTVAAKD CKLILTSKST LLTVEAFLTQ EQLKEESRFE RMGKQDDGKG DRSHKNEDGS ALASKQEMQY EITNYMVPAK NGTAGSSLFN EKEDSESDDS MHFDYSSNPN PKRQRCVV | |
Greater than 90% as determined by SDS-PAGE. |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |