missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA repair protein RAD51 homolog 4 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA repair protein RAD51 homolog 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6579-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001136043.1 | |
DNA repair protein RAD51 homolog 4 Protein | |
Research Use Only | |
RAD51L3 | |
Recombinant Protein | |
E. coli | |
MGVLRVGLCP GLTEEMIQLL RSHRIKTVVD LVSADLEEVA QKCGLSYKAL VALRRVLLAQ FSAFPVNGAD LYEELKTSTA ILSTGIGSLD KLLDAGLYTG EVTEIVGGPG SGKTQVCLCM AANVAHGLQQ NVLYVDSNGG LTASRLLQLL QAKTQDEEEQ AEALRRIQVV HAFDIFQMLD VLQELRGTVA QQVTGSSGTV KVVVVDSVTA VVSPLLGGQQ REGLALMMQL ARELKTLARD LGMAVVVTNH ITRDRDSGRL KPALGRSWSF VPSTRILLDT IEGAGASGGR RMACLAKSSR QPTGFQEMVD IGTWGTSEQS ATLQGDQT | |
Greater than 80% as determined by SDS-PAGE. |
5892 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |