missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Glutaminase liver isoform, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Glutaminase liver isoform, mitochondrial, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8210-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_001267727.1 | |
Glutaminase liver isoform, mitochondrial Protein | |
Research Use Only | |
GLS2 | |
Recombinant Protein | |
E. coli | |
GSHCGRGGWG HPSRSPLLGG GVRHHLSEAA AQGRETPHSH QPQHQDHDSS ESGMLSRLGD LLFYTIAEGQ ERIPIHKFTT ALKATGLQTS DPRLRDCMSE MHRVVQESSS GGLLDRDLFR KCVSSNIVLL TQAFRKKFVI PDFEEFTGHV DRIFEDVKEL TGGKVAAYIP QLAKSNPDLW GVSLCTVDGQ RHSVGHTKIP FCLQSCVKPL TYAISISTLG TDYVHKFVGK EPSGLRYNKL SLNEEGIPHN PMVNAGAIVV SSLIKMDCNK AEKFDFVLQY LNKMAGNEYM GFSNATFQSE KETGDRNYAI GYYLKEKKCF PKGVDMMAAL DLYFQLCSVE VTCESGSVMA ATLANGGICP ITGESVLSAE AVRNTLSLMH SCGMYDFSGQ FAFHVGLPAK SAVSGAILLV VPNVMGMMCL SPPLDKLGNS HRGTSFCQKL VSLFNFHNYD NLRHCARKLD PRREGAEIRN KTVVNLLFAA YSGDVSALRR FALSAMDMEQ KDYDSRTALH VAAAEGHIEV VKFLIEACKV NPFAKDRWGN IPLDDAVQFN HLEVVKLLQD YQDSYTLSET QAEAAAEALS KENLESMV | |
Greater than 90% as determined by SDS-PAGE. |
27165 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |