missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human GST-MDM2/HDM2 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

Brand:  R&D Systems™ E3-202-050

2219.38 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.


Product Code. 18128600

  • 2820.00 DKK / 50µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Recombinant Human GST-Mdm2 is a Ubiquitin ligase (E3) that functions downstream of a Ubiquitin-activating (E1) enzyme and a Ubiquitin-conjugating (E2) enzyme to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human GST-Mdm2 concentration of 0.5-4 μM.
Specifications

Specifications

Q00987-1
X mg/mL (X μM) in 50 mM Hepes pH 8, 200 mM NaCl, 10% glycerol, 5 mM TCEP
M.W. (theoretical): 82 kDa
50 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Unconjugated
>85%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Bioactivity
4193
GST MDM2/HDM2
E. coli-derived human MDM2/HDM2 protein Contains an N-terminal GST (glutathione S-transferase) tag
MDM2
Recombinant
MDM2/HDM2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers