missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Histone H2A.Z Protein
A DNA sequence encoding the Homo sapiens (Human) Histone H2A.Z, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6140-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_002097.1 | |
Histone H2A.Z Protein | |
Research Use Only | |
H2AFZ | |
Recombinant Protein | |
E. coli | |
AGGKAGKDSG KAKTKAVSRS QRAGLQFPVG RIHRHLKSRT TSHGRVGATA AVYSAAILEY LTAEVLELAG NASKDLKVKR ITPRHLQLAI RGDEELDSLI KATIAGGGVI PHIHKSLIGK KGQQKTV | |
Greater than 80% as determined by SDS-PAGE. |
3015 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |