missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Histone H2A.Z Protein

A DNA sequence encoding the Homo sapiens (Human) Histone H2A.Z, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6140-ec-1mg

 View more versions of this product

Product Code. 16091100

  • 15565.00 DKK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_002097.1
Histone H2A.Z Protein
Research Use Only
H2AFZ
Recombinant Protein
E. coli
AGGKAGKDSG KAKTKAVSRS QRAGLQFPVG RIHRHLKSRT TSHGRVGATA AVYSAAILEY LTAEVLELAG NASKDLKVKR ITPRHLQLAI RGDEELDSLI KATIAGGGVI PHIHKSLIGK KGQQKTV
Greater than 80% as determined by SDS-PAGE.
3015
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers