Learn More
R&D Systems™ Recombinant Human IL-4R alpha Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand:  R&D Systems™ 230-4R-025
2498.42 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18307447
  • 3265.00 DKK / 25µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Recombinant Human IL-4 R alpha Protein is derived from Sf 21 (baculovirus). The Recombinant Human IL-4 R alpha Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
24 kDa
25 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
3566
IL-4R alpha
Spodoptera frugiperda,Sf 21 (baculovirus)-derived human IL-4 R alpha protein Gly24-His232
CD124, CD124 antigen, IL-4 R alpha, IL-4 receptor subunit alpha, IL4R, IL4R alpha, IL-4R subunit alpha, IL-4RA, IL4RACD124, IL-4R-alpha, interleukin 4 receptor, interleukin-4 receptor alpha chain, interleukin-4 receptor subunit alpha
Recombinant
Product Suggestions