missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LAIR1 Protein
A cDNA sequence encoding the LAIR1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12526-20ug
This item is not returnable.
View return policy
Specifications
LAIR1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MQEEDLPRPS ISAEPGTVIP LGSHVTFVCR GPVGVQTFRL ERESRSTYND TEDVSQASPSESEARFRIDS VSEGNAGPYR CIYYKPPKWS EQSDYLELLV KETSG | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The protein (1 mg/ml) contains 20mM Tris-HCl (pH 7.5) 50mM NaCl and 1mM EDTA. |