missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFB10 Protein

A DNA sequence encoding the Homo sapiens (Human) NDUFB10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6413-ec-1mg

 View more versions of this product

Product Code. 16003140

  • 15565.00 DKK / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_004539.1
NDUFB10 Protein
Research Use Only
NDUFB10
Recombinant Protein
E. coli
MPDSWDKDVY PEPPRRTPVQ PNPIVYMMKA FDLIVDRPVT LVREFIERQH AKNRYYYYHR QYRRVPDITE CKEEDIMCMY EAEMQWKRDY KVDQEIINIM QDRLKACQQR EGQNYQQNCI KEVEQFTQVA KAYQDRYQDL GAYSSARKCL AKQRQRMLQE RKAAKEAAAA TS
Greater than 80% as determined by SDS-PAGE.
4716
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers