missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Netrin-4 Protein

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

4185.00 DKK

Specifications

Gene ID (Entrez) 59277
Molecular Weight (g/mol) 69 kDa
Name Netrin-4
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived human Netrin-4 protein Val26-Lys628,with a C-terminal 10-His tag
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18387707
View Documents
R&D Systems™
1254-N4-025/CF
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
4185.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
18127600
View Documents Promotion Details
R&D Systems™
1254-N4-025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
4185.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Human Netrin-4 Protein is derived from NS0. The Recombinant Human Netrin-4 Protein has been validated for the following applications: Binding Activity.
Specifications

Specifications

59277
Netrin-4
Mouse myeloma cell line,NS0-derived human Netrin-4 protein Val26-Lys628,with a C-terminal 10-His tag
Recombinant
69 kDa
25 μg
Unconjugated
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Videos
SDS
Documents

Documents

Product Certifications