missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NMNAT1 / NMNAT Protein

A DNA sequence encoding the Homo sapiens (Human) NMNAT1 / NMNAT, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8070-ec-200ug

 View more versions of this product

Product Code. 16092481

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001284707.1
NMNAT1 / NMNAT Protein
Research Use Only
NMNAT1
Recombinant Protein
E. coli
MENSEKTEVV LLACGSFNPI TNMHLRLFEL AKDYMNGTGR YTVVKGIISP VGDAYKKKGL IPAYHRVIMA ELATKNSKWV EVDTWESLQK EWKETLKVLR HHQEKLEASD CDHQQNSPTL ERPGRKRKWT ETQDSSQKKS LEPKTKAVPK VKLLCGADLL ESFAVPNLWK SEDITQIVAN YGLICVTRAG NDAQKFIYES DVLWKHRSNI HVVNEWIAND ISSTKIRRAL RRGQSIRYLV PDLVQEYIEK HNLYSSESED RNAGVILAPL QRNTAEAKT
Greater than 80% as determined by SDS-PAGE.
64802
200 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers