missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant Human Osteoprotegerin / TNFRSF11B Protein Product Code.: 15943818

enQuireBio™ Recombinant Human Osteoprotegerin / TNFRSF11B Protein

Product Code. 15943818
50 μg, 50µg
Click to view available options
Quantity:
10 μg
100 μg
250 μg
50 μg
Unit Size:
100µg
10µg
250µg
50µg
This item is not returnable. View return policy

Product Code. 15943818

Brand: enQuireBio™ QP10426ye50ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Human Osteoprotegerin / TNFRSF11B, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_002537.3
Gene ID (Entrez) 4982
Name Osteoprotegerin / TNFRSF11B Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than 1.0 EU/ug as determined by LAL method.
Gene Symbol TNFRSF11B
Protein Length Partial (see sequence information for more details).
Biological Activity The ED50 as determined by neutralizing the stimulation of U937 cells is less than 10 ng/ml, corresponding to a specific activity of > 1.0 M 105 IU/mg in the presence of 10 ng/mL soluble rHuRANKL (sRANKL).
Product Type Recombinant Protein
Cross Reactivity Human
Species Yeast
Protein Tag FC
Sequence ETFPPKYLHY DEETSHQLLC DKCPPGTYLK QHCTAKWKTV CAPCPDHYYT DSWHTSDECL YCSPVCKELQ YVKQECNRTH NRVCECKEGR YLEIEFCLKH RSCPPGFGVV QAGTPERNTV CKRCPDGFFS NETSSKAPCR KHTNCSVFGL LLTQKGNATH DNICSGNSES TQKCGIDVTL EPKSSDKTHT CPPCPAPEFE GAPSVFLFPP KPKDTLMISR TPEVTCVVVD VSHEDPEVKF NWYVDGVEVH NAKTKPREEQ YNSTYRVVSV LTVLHQDWLN GKEYKCKVSN KALPTPIEKT ISKAKGQPRE PQVYTLPPSR DELTKNQVSL TCLVKGFYPS DIAVEWESNG QPENNYKTTP PVLDSDGSFF LYSKLTVDKS RWQQGNVFSC SVMHEALHNH YTQKSLSLSP GK
Buffer 0.2um filtered 20 mM PBS, pH 6.0, 150 mM NaCl, 0.02 % Tween-80, lyophilized
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE and HPLC.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Human Osteoprotegerin / TNFRSF11B Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.