missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Paired box protein Pax-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Paired box protein Pax-2, was expressed in the hosts and tags indicated.
1185.00 DKK - 15565.00 DKK
Specifications
Accession Number | NP_000269.3 |
---|---|
Gene ID (Entrez) | 5076 |
Name | Paired box protein Pax-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056111
|
enQuireBio™
QP8719-ec-10ug |
10 μg | N/A | ||||||
16096111
|
enQuireBio™
QP8719-ec-50ug |
50 μg | N/A | ||||||
16046111
|
enQuireBio™
QP8719-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16076111
|
enQuireBio™
QP8719-ec-200ug |
200 μg | N/A | ||||||
16086111
|
enQuireBio™
QP8719-ec-500ug |
500 μg | N/A | ||||||
16066111
|
enQuireBio™
QP8719-ec-1mg |
1 mg |
15565.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_000269.3 | |
Paired box protein Pax-2 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5076 | |
Research Use Only | |
PAX2 | |
Recombinant Protein | |
E. coli | |
MDMHCKADPF SAMHPGHGGV NQLGGVFVNG RPLPDVVRQR IVELAHQGVR PCDISRQLRV SHGCVSKILG RYYETGSIKP GVIGGSKPKV ATPKVVDKIA EYKRQNPTMF AWEIRDRLLA EGICDNDTVP SVSSINRIIR TKVQQPFHPT PDGAGTGVTA PGHTIVPSTA SPPVSSASND PVGSYSINGI LGIPRSNGEK RKRDEVEVYT DPAHIRGGGG LHLVWTLRDV SEGSVPNGDS QSGVDSLRKH LRADTFTQQQ LEALDRVFER PSYPDVFQAS EHIKSEQGNE YSLPALTPGL DEVKSSLSAS TNPELGSNVS GTQTYPVVTG RDMASTTLPG YPPHVPPTGQ GSYPTSTLAG MVP | |
Greater than 90% as determined by SDS-PAGE. |