missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human PAK3 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00005063-Q01-25ug
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
5063 | |
25 μg | |
beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
Protein Kinase | |
PAK3 |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
35.53 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
PAK3 | |
>80% by SDS-PAGE and Coomassie blue staining |