missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Reelin Protein
A DNA sequence encoding the Homo sapiens (Human) Reelin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6597-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_005036.2 | |
Reelin Protein | |
Research Use Only | |
RELN | |
Recombinant Protein | |
E. coli | |
AAGYYPRFSP FFFLCTHHGE LEGDGEQGEV LISLHIAGNP TYYVPGQEYH VTISTSTFFD GLLVTGLYTS TSVQASQSIG GSSAFGFGIM SDHQFGNQFM CSVVASHVSH LPTTNLSFIW IAPPAGTGCV NFMATATHRG QVIFKDALAQ QLCEQGAPTD VTVHPHLAEI HSDSIILRDD FDSYHQLQLN PNIWVECNNC ETGEQCGAIM HGNAVTFCEP YGPRELITT | |
Greater than 90% as determined by SDS-PAGE. |
5649 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |