missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
R&D Systems™ Recombinant Human SIRP gamma/CD172g His Avi-tag Protein, CF
Beskrivelse
Measured by its binding ability in a functional ELISA. Biotinylated Recombinant Human SIRP gamma/CD172g His-tag Avi-tag (Catalog # AVI9999) binds Recombinant Human CD47 Fc Chimera (Catalog # 4670-CD ) with an ED50 of 0.150-3.00 μg/mL.
Tekniske data
Tekniske data
| Adgangsnummer | Q9P1W8.3 |
| Til brug med (applikation) | Bioactivity |
| Formulering | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gen-id (Entrez) | 55423 |
| Molekylvægt (g/mol) | MolecularWeight-observed: 42-49 kDa, under reducing conditions., MolecularWeight-theroretical: 39 kDa |
| Mængde | 50 μg |
| Kilde | Chinese Hamster Ovary cell line, CHO-derived human SIRP gamma/CD172g protein Human SIRPG (Glu29-Pro360) (N-terminus) Accession # Q9P1W8.3 6-His tag Avi-tag (C-terminus) |
| Opbevaringskrav | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution. |
| Endotoksin koncentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | bA77C3.1, CD172g, CD172g antigen, FLJ42230, signal-regulatory protein beta 2, Signal-regulatory protein beta-2, signal-regulatory protein gamma, SIRP beta 2, SIRP-b2, SIRPB2CD172 antigen-like family member B, SIRP-beta-2, SIRPG, SIRPgamma, SIRP-gamma |
| Vis mere |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?