Learn More
R&D Systems™ Recombinant Human Sphingosine Kinase 1/SPHK1 Protein Product Code.: 18187580
R&D Systems™ Recombinant Human Sphingosine Kinase 1/SPHK1 Protein
Product Code. 18187580
10ug; Carrier Free
Click to view available options
:
10ug; Carrier Free
Product Code. 18187580
Brand: R&D Systems™ 5536SK010
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein is derived from Sf 21 (baculovirus). The Recombinant Human Sphingosine Kinase 1/SPHK1 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl, Glycerol, DTT and Triton™ .
Gene ID (Entrez) 8877
Molecular Weight (g/mol) 45 kDa
Name Sphingosine Kinase 1/SPHK1
Quantity 10ÎĽg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human Sphingosine Kinase 1/SPHK1 protein Asp2-Leu398 with an N-terminal Met and 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >75%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Product Content Correction
Product Title
R&D Systems™ Recombinant Human Sphingosine Kinase 1/SPHK1 Protein >