missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Spliceosome RNA helicase DDX39B Protein
A DNA sequence encoding the Homo sapiens (Human) Spliceosome RNA helicase DDX39B, was expressed in the hosts and tags indicated.
1185.00 DKK - 13770.00 DKK
Specifications
Accession Number | NP_004631.1 |
---|---|
Gene ID (Entrez) | 7919 |
Name | Spliceosome RNA helicase DDX39B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063961
|
enQuireBio™
QP8315-ec-10ug |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003971
|
enQuireBio™
QP8315-ec-50ug |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16053961
|
enQuireBio™
QP8315-ec-100ug |
100 μg |
3450.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16083961
|
enQuireBio™
QP8315-ec-200ug |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093961
|
enQuireBio™
QP8315-ec-500ug |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073961
|
enQuireBio™
QP8315-ec-1mg |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004631.1 | |
Spliceosome RNA helicase DDX39B Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
7919 | |
Research Use Only | |
DDX39B | |
Recombinant Protein | |
E. coli | |
AENDVDNELL DYEDDEVETA AGGDGAEAPA KKDVKGSYVS IHSSGFRDFL LKPELLRAIV DCGFEHPSEV QHECIPQAIL GMDVLCQAKS GMGKTAVFVL ATLQQLEPVT GQVSVLVMCH TRELAFQISK EYERFSKYMP NVKVAVFFGG LSIKKDEEVL KKNCPHIVVG TPGRILALAR NKSLNLKHIK HFILDECDKM LEQLDMRRDV QEIFRMTPHE KQVMMFSATL SKEIRPVCRK FMQDPMEIFV | |
Greater than 90% as determined by SDS-PAGE. |