missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human Talin1 GST (N-Term) Protein

Recombinant Protein

Brand:  Novus Biologicals™ H00007094-Q06-25ug

 View more versions of this product

Product Code. 18310898

  • 5966.37 DKK / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications

Specifications

Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification
7094
25 μg
ILWEQ, KIAA1027, talin 1, talin-1, TLN
Cell Biology, Cellular Markers
Talin1
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
173.58 kDa
Store at -80°C. Avoid freeze-thaw cycles.
TLN1
>80% by SDS-PAGE and Coomassie blue staining
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers