missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TC-PTP (aa 2-314) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 1930-PT-050
2021.54 DKK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Measured by its ability to dephosphorylate a tyrosine residue in a peptide containing the EGFR Y992 phosphorylation site (Catalog # ES006 ). The specific activity is >30 μmol/min/mg, as measured under the described conditions.Specifications
Q96AU5 | |
Supplied as a 0.2 μm filtered solution in HEPES, NaCl, Glycerol and Betamercaptoethanol. | |
M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 37 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
PTPN2 | |
Recombinant | |
TC-PTP/PTPN2 |
Enzyme Activity | |
5771 | |
TC-PTP aa 2-314 | |
E. coli-derived human TC-PTP protein Thr2-Asn314, with an N-terminal Met and 6-His tag | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain |