Learn More
R&D Systems™ Recombinant Human TSG-6 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
Brand: R&D Systems™ 2104-TS-050
This item is not returnable.
View return policy
Description
The Recombinant Human TSG-6 Protein is derived from NS0. The Recombinant Human TSG-6 Protein has been validated for the following applications: Binding Activity.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
30.5 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
7130 | |
TSG-6 | |
Mouse myeloma cell line,NS0-derived human TSG-6 protein Trp18-Leu277,with a C-terminal 10-His tag | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Product Title
R&D Systems™ Recombinant Human TSG-6 Protein > 50ug; Carrier Free
Spot an opportunity for improvement?Share a Content Correction