missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Zinc finger protein 175 Protein
A DNA sequence encoding the Homo sapiens (Human) Zinc finger protein 175, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6904-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_009078.1 | |
Zinc finger protein 175 Protein | |
Research Use Only | |
ZNF175 | |
Recombinant Protein | |
E. coli | |
MPADVNLSQK PQVLGPEKQD GSCEASVSFE DVTVDFSREE WQQLDPAQRC LYRDVMLELY SHLFAVGYHI PNPEVIFRML KEKEPRVEEA EVSHQRCQER EFGLEIPQKE ISKKASFQKD MVGEFTRDGS WCSILEELRL DADRTKKDEQ NQIQPMSHSA FFNKKTLNTE SNCEYKDPGK MIRTRPHLAS SQKQPQKCCL FTESLKLNLE VNGQNESNDT EQLDDVVGSG QLFSHSSSDA CSKNIHTGET FCKGNQCRKV CGHKQSLKQH QIHTQKKPDG CSECGGSFTQ KSHLFAQQRI HSVGNLHECG KCGKAFMPQL KLSVYLTDHT GDIPCICKEC GKVFIQRSEL LTHQKTHTRK KPYKCHDCGK AFFQMLSLFR HQRTHSREKL YECSECGKGF SQNSTLIIHQ KIHTGERQYA CSECGKAFTQ KSTLSLHQRI HSGQKSYVCI ECGQAFIQKA HLIVHQRSHT GEKPYQCHNC GKSFISKSQL DIHHRIHTGE KPYECSDCGK TFTQKSHLNI HQKIHTGERH HVCSECGKAF NQKSILSMHQ RIHTGEKPYK CSECGKAFTS KSQFKEHQRI HTGEKPYVCT ECGKAFNGRS NFHKHQITHT RERPFVCYKC GKAFVQKSEL ITHQRTHMGE KPYECLDCGK SFSKKPQLKV HQRIHTGERP YVCSECGKAF NNRSNFNKHQ TTHTRDKSYK CSYSVKGFTK Q | |
Greater than 90% as determined by SDS-PAGE. |
7728 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |