missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse BMP-4 Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

4890.00 DKK

Specifications

Gene ID (Entrez) 12159
Molecular Weight (g/mol) Predicted MW: 13.1kDa (monomer) Observed MW: 18-26kDa, reducing conditions
Name BMP-4
Quantity 10 μg
Source Chinese Hamster Ovary cell line,CHO-derived mouse BMP-4 protein Ser293-Arg408
View More Specs

Products 2
Product Code Brand Formulation Price Quantity & Availability  
Product Code Brand Formulation Price Quantity & Availability  
18157000
View Documents
R&D Systems™
5020-BP-010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein.
4890.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
18128490
View Documents Promotion Details
R&D Systems™
5020-BP-010/CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
4890.00 DKK
10µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Recombinant Mouse BMP-4 Protein is derived from CHO. The Recombinant Mouse BMP-4 Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

12159
BMP-4
Chinese Hamster Ovary cell line,CHO-derived mouse BMP-4 protein Ser293-Arg408
Disulfide-linked homodimer
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Predicted MW: 13.1kDa (monomer) Observed MW: 18-26kDa, reducing conditions
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
BMP2B, BMP-2B, BMP2B1, BMP2BMCOPS6, BMP4, BMP-4, Bone morphogenetic protein 2B, bone morphogenetic protein 4, DVR4, MCOPS6, OFC11, ZYME
Recombinant
Videos
SDS
Documents

Documents

Product Certifications