Learn More
R&D Systems™ Recombinant Mouse FGF-4 (aa 67-202) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
Brand: R&D Systems™ 7486-F4-025/CF
This item is not returnable.
View return policy
Description
The Recombinant Mouse FGF-4 (aa 67-202) Protein is derived from E. coli. The Recombinant Mouse FGF-4 (aa 67-202) Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and EDTA. | |
15.2 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
14175 | |
FGF-4 (aa 67-202) | |
E. coli-derived mouse FGF-4 protein Ser67-Leu202 | |
Monomer | |
Recombinant |
Product Content Correction