missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse IL-34 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
4035.00 DKK
Specifications
Gene ID (Entrez) | 76527 |
---|---|
Molecular Weight (g/mol) | 25.4 kDa |
Name | IL-34 |
Quantity | 10 μg |
Source | Mouse myeloma cell line,NS0-derived mouse IL-34 protein Asn21-Pro235,with a C-terminal 6-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18168170
|
R&D Systems™
5195-ML-010 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
4035.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18167670
|
R&D Systems™
5195-ML-010/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
4035.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Mouse IL-34 Protein is derived from NS0. The Recombinant Mouse IL-34 Protein has been validated for the following applications: Bioactivity.Specifications
76527 | |
IL-34 | |
Mouse myeloma cell line,NS0-derived mouse IL-34 protein Asn21-Pro235,with a C-terminal 6-His tag | |
Noncovalently-linked homodimer | |
Recombinant |
25.4 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |