missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Nqo2 Protein
A DNA sequence encoding the Mus musculus (Mouse) Nqo2, was expressed in the hosts and tags indicated.
1820.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_001156713.1 |
---|---|
Gene ID (Entrez) | 18105 |
Name | Nqo2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083071
|
enQuireBio™
QP8176-ec-10ug |
10 μg | N/A | ||||||
16023081
|
enQuireBio™
QP8176-ec-50ug |
50 μg | N/A | ||||||
16073071
|
enQuireBio™
QP8176-ec-100ug |
100 μg |
5015.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16003081
|
enQuireBio™
QP8176-ec-200ug |
200 μg | N/A | ||||||
16013081
|
enQuireBio™
QP8176-ec-500ug |
500 μg | N/A | ||||||
16093071
|
enQuireBio™
QP8176-ec-1mg |
1 mg |
22245.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001156713.1 | |
Nqo2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
18105 | |
Research Use Only | |
Nqo2 | |
Recombinant Protein | |
E. coli | |
MAGKKVLIVY AHQEPKSFNG SLKKVAVEEL SKQGCTVTVS DLYSMNFEPR ATRNDITGAP SNPDVFSYGI ETHEAYKKKA LTSDIFEEQR KVQEADLVIF QFPLYWFSVP AILKGWMDRV LCRGFAFDIP GFYDSGFLKG KLALLSLTTG GTAEMYTKDG VSGDFRYFLW PLQHGTLHFC GFKVLAPQIS FGLDVSSEEE RKVMLASWAQ RLKSIWKEEP IHCTPPWYFQ E | |
Greater than 90% as determined by SDS-PAGE. |