missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Nuclear receptor ROR-gamma Protein

A DNA sequence encoding the Mus musculus (Mouse) Nuclear receptor ROR-gamma, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9374-ye-50ug

 View more versions of this product

Product Code. 16078201

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001280663.1
Nuclear receptor ROR-gamma Protein
Research Use Only
RORC
Recombinant Protein
Yeast
MDRAPQRHHR TSRELLAAKK THTSQIEVIP CKICGDKSSG IHYGVITCEG CKGFFRRSQQ CNVAYSCTRQ QNCPIDRTSR NRCQHCRLQK CLALGMSRDA VKFGRMSKKQ RDSLHAEVQK QLQQQQQQEQ VAKTPPAGSR GADTLTYTLG LSDGQLPLGA SPDLPEASAC PPGLLRASGS GPPYSNTLAK TEVQGASCHL EYSPERGKAE GRDSIYSTDG QLTLGRCGLR FEETRHPELG EPEQGPDSHC IPSFCSAPEV PYASLTDIEY LVQNVCKSFR ETCQLRLEDL LRQRTNLFSR EEVTSYQRKS MWEMWERCAH HLTEAIQYVV EFAKRLSGFM ELCQNDQIIL LTAGAMEVVL VRMCRAYNAN NHTVFFEGKY GGVELFRALG CSELISSIFD FSHFLSALCF SEDEIALYTA LVLINANRPG LQEKRRVEHL QYNLELAFHH HLCKTHRQGL LAKLPPKGKL RSLCSQHVEK LQIFQHLHPI VVQAAFPPLY KELFSTDVES PEGLSK
Greater than 90% as determined by SDS-PAGE.
19885
50 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions