missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Protein delta homolog 2 Protein

A DNA sequence encoding the Mus musculus (Mouse) Protein delta homolog 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9838-ye-100ug

 View more versions of this product

Product Code. 16078831

  • 6005.00 DKK / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_001272942.1
Protein delta homolog 2 Protein
Research Use Only
DLK2
Recombinant Protein
Yeast
DDCSSHCDLA HGCCAPDGSC RCDPGWEGLH CERCVRMPGC QHGTCHQPWQ CICHSGWAGK FCDKDEHICT SQSPCQNGGQ CVYDGGGEYH CVCLPGFHGR GCERKAGPCE QAGFPCRNGG QCQDNQGFAL NFTCRCLAGF MGAHCEVNVD DCLMRPCANG ATCIDGINRF SCLCPEGFAG RFCTINLDDC ASRPCQRGAR CRDRVHDFDC LCPSGYGGKT CELVLPAPEP ASVGTPQMPT SAVVVPATGP APHSAGAGLL RISVKEVVRR QESGLGESS
Greater than 90% as determined by SDS-PAGE.
106565
100 μg
Not determined.
Extracellular Domain
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers