missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Tyrosine-protein kinase JAK1 Protein
A DNA sequence encoding the Mus musculus (Mouse) Tyrosine-protein kinase JAK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6245-ec-100ug
This item is not returnable.
View return policy
Specifications
Tyrosine-protein kinase JAK1 Protein | |
Research Use Only | |
JAK1 | |
Recombinant Protein | |
E. coli | |
EEQNPDIVSE KQPTTEVDPT HFEKRFLKRI RDLGEGHFGK VELCRYDPEG DNTGEQVAVK SLKPESGGNH IADLKKEIEI LRNLYHENIV KYKGICMEDG GNGIKLIMEF LPSGSLKEYL PKNKNKINLK QQLKYAIQIC KGMDYLGSRQ YVHRDLAARN VLVESEHQVK IGDFGLTKAI ETDKEYYTVK DDRDSPVFWY APECLIQCKF YIASDVWSFG VTLHELLTYC DSDFSPMALF LKMIGPTHGQ MTVTRLVKTL KEGKRLPCPP NCPDEVYQLM RKCWEFQPSN RTTFQNLIEG FEALL | |
Greater than 90% as determined by SDS-PAGE. |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction