missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa PA-I galactophilic lectin Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) PA-I galactophilic lectin, was expressed in the hosts and tags indicated. option, ab
Brand: enQuireBio™ QP6995-1mg
This item is not returnable.
View return policy
Specifications
NP_251260.1 | |
P. aeruginosa PA-I galactophilic lectin Protein | |
Research Use Only | |
lecA | |
Recombinant Protein | |
E. coli | |
AWKGEVLANN EAGQVTSIIY NPGDVITIVA AGWASYGPTQ KWGPQGDREH PDQGLICHDA FCGALVMKIG NSGTIPVNTG LFRWVAPNNV QGAITLIYND VPGTYGNNSG SFSVNIGKDQ S | |
Greater than 90% as determined by SDS-PAGE. |
882335 | |
1 mg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His | |
Tris-based buffer, 50% glycerol |