missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit VEGF / VEGFA / VEGF165 Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) VEGF / VEGFA / VEGF165, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6879-ye-10ug
This item is not returnable.
View return policy
Specifications
NP_999249.1 | |
VEGF / VEGFA / VEGF165 Protein | |
Research Use Only | |
VEGFA | |
Recombinant Protein | |
Yeast | |
YLWLLHAPLL HVSLDSLVAA FSVVFEVLPS SLDIPGSKKE EQASTQLGGV EEEHEASGVV TRQLAFVVDA ITNPTLEKET KTITRIHKAP KFPSHFGFWK RAEEERGGKS SGEKSRKTDG VGERAQAGEQ REGPGQRAAA LTDRQTDTAP SPSAHLLPGR RPTVDAAASG GQQPEPAPEG GVEGVGARGI ALKLFVQLLG CSRSGGVVVR AGGAEPSGAA RSVSSGREEP PPPPPEEEGE EEGEKEEERG PRWRLGAGEP GSWTGEAAVC ADSAPAARAP QALARASAPG ARGARGGAEE SGPSRSPSRR GSASRAGPGR ASETMNFLLS WVHWSLALLL YLHHAKWSQA APMAEEGDNK PHEVVKFMEV YRRSYCQPIE TLVDIFQEYP DEIEYIFKPS CVPLVRCGGC CNDESLECVP TEEFNVTMQI MRIKPHQGQH IGEMSFLQHN KCECRCDKPR R | |
Greater than 90% as determined by SDS-PAGE. |
397157 | |
10 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |