missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat HGF / Hepatocyte Growth Factor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) HGF / Hepatocyte Growth Factor, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8607-ec-200ug
This item is not returnable.
View return policy
Specifications
NP_058713.1 | |
HGF / Hepatocyte Growth Factor Protein | |
Research Use Only | |
HGF | |
Recombinant Protein | |
E. coli | |
TIVNLDHPVI SCAKTKQLRV VNGIPTQTTV GWMVSLKYRN KHICGGSLIK ESWVLTARQC FPARNKDLKD YEAWLGIHDV HERGEEKRKQ ILNISQLVYG PEGSDLVLLK LARPAILDNF VSTIDLPSYG CTIPEKTTCS IYGWGYTGLI NADGLLRVAH LYIMGNEKCS QHHQGKVTLN ESELCAGAEK IGSGPCEGDY GGPLICEQHK MRMVLGVIVP GRGCAIPNRP GIFVRVAYYA KWIHKVILTY KL | |
Greater than 90% as determined by SDS-PAGE. |
24446 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |