missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6780-ec-500ug

 View more versions of this product

Product Code. 16095790

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_067589.1
TGF-beta 1 / TGFB1 Protein
Research Use Only
TGFB1
Recombinant Protein
E. coli
LSTCKTIDME LVKRKRIEAI RGQILSKLRL ASPPSQGEVP PGPLPEAVLA LYNSTRDRVA GESADPEPEP EADYYAKEVT RVLMVDRNNA IYDKTKDITH SIYMFFNTSD IREAVPEPPL LSRAELRLQR FKSTVEQHVE LYQKYSNNSW RYLGNRLLTP TDTPEWLSFD VTGVVRQWLN QGDGIQGFRF SAHCSCDSKD NVLHVEINGI SPKRRGDLGT IHDMNRPFLL LMATPLERAQ HLHSSRHRR
Greater than 80% as determined by SDS-PAGE.
59086
500 μg
Not determined.
Partial (see sequence information for more details).
Rat
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers