missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Vasopressin V1a receptor Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Vasopressin V1a receptor, was expressed in the hosts and tags indicated.
1820.00 DKK - 22245.00 DKK
Specifications
Accession Number | NP_444178.2 |
---|---|
Gene ID (Entrez) | 25107 |
Name | Vasopressin V1a receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046781
|
enQuireBio™
QP8843-ec-10ug |
10 μg | N/A | ||||||
16086781
|
enQuireBio™
QP8843-ec-50ug |
50 μg | N/A | ||||||
16036781
|
enQuireBio™
QP8843-ec-100ug |
100 μg |
5015.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16066781
|
enQuireBio™
QP8843-ec-200ug |
200 μg | N/A | ||||||
16076781
|
enQuireBio™
QP8843-ec-500ug |
500 μg | N/A | ||||||
16056781
|
enQuireBio™
QP8843-ec-1mg |
1 mg |
22245.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_444178.2 | |
Vasopressin V1a receptor Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
GST | |
Tris-based buffer, 50% glycerol |
25107 | |
Research Use Only | |
Avpr1a | |
Recombinant Protein | |
E. coli | |
SQDRSVGNSS PWWPLTTEGS NGSQEAARLG EGDSPLGDVR NEELAK | |
Greater than 90% as determined by SDS-PAGE. |