missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Virus Chikungunya Virus E2 Fc Protein
3975.00 DKK - 14100.00 DKK
Specifications
Concentration | 0.32 mg/ml |
---|---|
For Use With (Application) | ELISA, SDS-PAGE |
Formulation | 20mM HEPES, pH7.0, 300mM NaCl, 0.25% Triton X-100 |
Molecular Weight (g/mol) | theroretical M.W.: 69 kDa |
Storage Requirements | Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18334582
|
Novus Biologicals
NBP3-14818-0.1MG |
0.1 mg |
3975.00 DKK
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18325714
|
Novus Biologicals
NBP3-14818-0.5MG |
0.5 mg |
14100.00 DKK
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
0.32 mg/ml | |
20mM HEPES, pH7.0, 300mM NaCl, 0.25% Triton X-100 | |
Store at 4C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. | |
>95% |
ELISA, SDS-PAGE | |
theroretical M.W.: 69 kDa | |
Chikungunya, E2 envelope glycoprotein, P130, Spike glycoprotein E2 | |
Chikungunya Virus E2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title