Learn More
Kautex™ Red Polypropylene Screw Closures
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
Brand:  Kautex™ 2000084198
 View more versions of this product
Product Code. 10196380
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • PP
  • Red
  • Tamper evident ring
  • PE-foam insert
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Polypropylene
Screw Cap
28 mm
Tamper Evident Ring and Polyethylene Foam Insert
Polyethylene Foam Insert
50 mL Bottle