Learn More
Kautex™ Red Polypropylene Screw Closures
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
Brand: Kautex™ 2000084006
This item is not returnable.
View return policy
Description
- PP
- Red
- Tamper evident ring
- PE-foam insert
Specifications
Red | |
Polypropylene | |
Screw Cap | |
32 mm |
Pull-off Ring and Polyethylene Foam Insert | |
Polyethylene Foam Insert | |
10 to 1000 mL Bottle |