Learn More
BINDER™ Reinforced Inner Chamber
Reinforced inner chamber for climate chambers and cooling incubation chambers.
Brand:  BINDER™ 8012-2521
 View more versions of this product
Product Code. 30238263
  • 8979.78 DKK / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Reinforced inner chamber for bulky and heavy samples.
  • Weight capacity ranged from 100 to 200 kg, depending on model.
Specifications
Specifications
KBF 720
Stainless Steel
Chamber Rack
200 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BINDER™ Reinforced Inner Chamber > For use with KBF 720