Learn More
Memmert™ Reinforced Stainless Steel Shelf
Additional reinforced stainless steel shelf for reinforced chamber option
Brand: Memmert™ IPP750ecoB32191
This item is not returnable.
View return policy
Description
- With guide bars and fixing screws
- Requires option K1
- Consider max. loading of chamber; max. loading 60 kg
Specifications
IPP750eco | |
Stainless Steel |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ Reinforced Stainless Steel Shelf > IPP750eco
Spot an opportunity for improvement?Share a Content Correction