Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20410
587.88 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10026732
  • 653.00 DKK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard 0.5
15 x 103 mm
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6226138 Certificate TypeCertificate of Analysis Date06/05/2025 Catalog NumberR20410
Lot Number6212741 Certificate TypeCertificate of Analysis Date02/04/2025 Catalog NumberR20410
Lot Number6212740 Certificate TypeCertificate of Analysis Date03/03/2025 Catalog NumberR20410
Lot Number6185618 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR20410
Lot Number6185617 Certificate TypeCertificate of Analysis Date28/11/2024 Catalog NumberR20410
Promotions
Promotions
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
Product Content Correction
Product Title
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards > Turbidity Standard 0.5