Learn More
Thermo Scientific™ Remel™ Streptex™ Extraction Enzyme
Remel Streptex and Extraction Enzyme are used with Streptex kit.
Brand: Thermo Scientific™ R30951001
376.45 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Streptex™ supplementary reagents are intended for use in the streptococcal grouping system provided by the Streptex kit, which contains all components necessary for the system. These reagents are provided as replacement items. For complete information and correct use of this product, the user is referred to the Instructions for Use which accompanies the Streptex kits (ZL50/R30950501 and ZL61/R30164701).
Ordering Alerts:
Freeze dried.
Specifications
Streptex and Extraction Enzyme | |
Streptococcus Test Kit | |
11 mL/Vial, 25 Tests/Kit |
Yes | |
25 Tests/Kit |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6154645 | Certificate of Analysis | 06/01/2025 | R30951001 |
6129042 | Certificate of Analysis | 28/11/2024 | C22322.30 |
6003560 | Certificate of Analysis | 18/07/2024 | R30951001 |
6003379 | Certificate of Analysis | 15/07/2024 | R30951001 |
3729830 | Certificate of Analysis | 24/01/2024 |