Learn More
Labconco™ Replacement Connecting Bulbs
These Replacement Connecting Bulbs separate liquid from vapors. Designed for Kjeldahl Units.
Brand:  Labconco™ 2081300
Product Code. 12770087
  • 4035.00 DKK / Pack of 6
Explore more special offers
Description
Description
  • The connecting bulb separates liquid from vapors prior to the vapors entering the condenser.
  • Double baffle design effectively traps caustic mist that may carry over to the recovery solution.
  • Provide connection between a Kjeldahl flask and distillation rack.
  • One-piece borosilicate glass.
  • Each piece of glassware may be labeled 20813-01.
Specifications
Specifications
Connecting Bulb
1.8 kg
Kjeldahl Units
Borosilicate Glass
4 lb.
6 Pk.