Learn More
Thermo Scientific Replacement Rubber Bands
Replacement Rubber Bands (Set of 5).
Brand: Thermo Scientific 75003030
236.49 DKK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
- Replacement Rubber Bands (Set of 5)
Specifications
Rubber Bands | |
Centrifuge | |
5 Pk. |
Rubber | |
Reusable |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%