Learn More
Cole-Parmer™ Respirator Storage Bag
Shop All Cole-Parmer Products
Click to view available options
:
Height: 381mm
Description
- Cleaning instructions are printed right on the bag for easy reference.
- Measure 12 x 15 in.
Specifications
Specifications
Product Type | Carrying Case |
Color | Transparent |
For Use With (Application) | Protects The Mask of Dust and Contamination |
Height (Metric) | 381mm |
Product Content Correction
Product Title
Cole-Parmer™ Respirator Storage Bag >
Spot an opportunity for improvement?Share a Content Correction